Skip to Content
MilliporeSigma
2-Hydroxy-3-(1H-1,2,4-triazol-1-yl)propanoic acid TraceCERT®, certified reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

2-Hydroxy-3-(1H-1,2,4-triazol-1-yl)propanoic acid

2-Hydroxy-3-(1H-1,2,4-triazol-1-yl)propanoic acid TraceCERT®, certified reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
α-Hydroxy-1H-1,2,4-triazole-1-propanoic acid
Empirical Formula (Hill Notation):
C5H7N3O3
CAS Number:
Molecular Weight:
157.13

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade