Skip to Content
MilliporeSigma
2-Ethylhexanoic acid Pharmaceutical Secondary Standard; Certified Reference Material

2-Ethylhexanoic acid

2-Ethylhexanoic acid Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2-Ethylcaproic acid, 2-Ethylhexanoic acid
Linear Formula:
CH3(CH2)3CH(C2H5)CO2H
CAS Number:
Molecular Weight:
144.21
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade
Technique