Skip to Content
MilliporeSigma
2-Chlorobenzoyl chloride 95%

2-Chlorobenzoyl chloride

2-Chlorobenzoyl chloride 95%
Synonym(s):
2-Chlorobenzoyl chloride
Linear Formula:
2-(Cl)C6H4COCl
CAS Number:
Molecular Weight:
175.01

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing