Skip to Content
MilliporeSigma
2-Chloro-2-propen-1-ol technical grade, 90%

2-Chloro-2-propen-1-ol

2-Chloro-2-propen-1-ol technical grade, 90%
Synonym(s):
2-Chloroallyl alcohol
Linear Formula:
H2C=C(Cl)CH2OH
CAS Number:
Molecular Weight:
92.52

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing