Skip to Content
MilliporeSigma
2-(Benzylthio)-3-nitropyridine 98%

2-(Benzylthio)-3-nitropyridine

2-(Benzylthio)-3-nitropyridine 98%
Empirical Formula (Hill Notation):
C12H10N2O2S
CAS Number:
Molecular Weight:
246.29

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing