Skip to Content
MilliporeSigma
2-Amino-5-nitrobenzonitrile 95%

2-Amino-5-nitrobenzonitrile

2-Amino-5-nitrobenzonitrile 95%
Synonym(s):
2-Cyano-4-nitroaniline, 5-Nitroanthranilonitrile
Linear Formula:
O2NC6H3(NH2)CN
CAS Number:
Molecular Weight:
163.13
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing