Skip to Content
MilliporeSigma
2-Amino-4-chlorobenzonitrile 99%

2-Amino-4-chlorobenzonitrile

2-Amino-4-chlorobenzonitrile 99%
Synonym(s):
4-Chloroanthranilonitrile, 5-Chloro-2-cyanoaniline
Linear Formula:
H2NC6H3(Cl)CN
CAS Number:
Molecular Weight:
152.58
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing