Skip to Content
MilliporeSigma
2-Amino-3-hydroxypyridine analytical standard

2-Amino-3-hydroxypyridine

2-Amino-3-hydroxypyridine analytical standard
Synonym(s):
2-Amino-3-pyridinol
Empirical Formula (Hill Notation):
C5H6N2O
CAS Number:
Molecular Weight:
110.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing