Skip to Content
MilliporeSigma
2-Acetylpyrazine ≥99%, FCC, FG

2-Acetylpyrazine

2-Acetylpyrazine ≥99%, FCC, FG
Synonym(s):
Acetylpyrazine
Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing