Skip to Content
MilliporeSigma
2,6-Lutidine analytical standard

2,6-Lutidine

2,6-Lutidine analytical standard
Synonym(s):
2,6-Dimethylpyridine
Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing