Skip to Content
MilliporeSigma
2,6-Diisopropylaniline technical grade, 90%

2,6-Diisopropylaniline

2,6-Diisopropylaniline technical grade, 90%
Linear Formula:
[(CH3)2CH]2C6H3NH2
CAS Number:
Molecular Weight:
177.29
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing