Skip to Content
MilliporeSigma
2,5-Dihydroxybenzoic acid analytical standard

2,5-Dihydroxybenzoic acid

2,5-Dihydroxybenzoic acid analytical standard
Synonym(s):
2,5-DHBA, DHB, Gentisic acid, Hydroquinonecarboxylic acid, 2,5-Dihydroxybenzoic acid
Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Analyte Suitability
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique