Skip to Content
MilliporeSigma
2,5-Bis(trifluoromethyl)aniline 99%

2,5-Bis(trifluoromethyl)aniline

2,5-Bis(trifluoromethyl)aniline 99%
Linear Formula:
(CF3)2C6H3NH2
CAS Number:
Molecular Weight:
229.12

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing