Skip to Content
MilliporeSigma
2-(4-Hydroxyphenyl)ethanol 98%

2-(4-Hydroxyphenyl)ethanol

2-(4-Hydroxyphenyl)ethanol 98%
Synonym(s):
4-Hydroxyphenethyl alcohol, Tyrosol, p-HPEA, 2-(4-Hydroxyphenyl)ethanol
Linear Formula:
HOC6H4CH2CH2OH
CAS Number:
Molecular Weight:
138.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing