Skip to Content
MilliporeSigma
2,3-Dihydroxyquinoxaline 98%

2,3-Dihydroxyquinoxaline

2,3-Dihydroxyquinoxaline 98%
Synonym(s):
2,3(1H,4H)-Quinoxalinedione, 2,3-Quinoxalinediol
Empirical Formula (Hill Notation):
C8H6N2O2
CAS Number:
Molecular Weight:
162.15
EC Number:

Products

Product Category
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing