Skip to Content
MilliporeSigma
2,2′-Thiodiacetic acid 98%

2,2′-Thiodiacetic acid

2,2′-Thiodiacetic acid 98%
Synonym(s):
2,2′-Thio-bis(acetic acid), Dicarboxydimethyl sulfide, Thiodiglycolic acid, 2,2′-Thiodiacetic acid
Linear Formula:
S(CH2COOH)2
CAS Number:
Molecular Weight:
150.15

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing