Skip to Content
MilliporeSigma
2,2′-(Ethylenedioxy)diethanethiol 95%

2,2′-(Ethylenedioxy)diethanethiol

2,2′-(Ethylenedioxy)diethanethiol 95%
Synonym(s):
1,2-Bis(2-mercaptoethoxy)ethane, 3,6-Dioxa-1,8-octane-dithiol
Linear Formula:
HSCH2CH2OCH2CH2OCH2CH2SH
CAS Number:
Molecular Weight:
182.30
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing