Skip to Content
MilliporeSigma
1-Z-4-Piperidone 99%

1-Z-4-Piperidone

1-Z-4-Piperidone 99%
Synonym(s):
1-(Benzyloxycarbonyl)-4-piperidinone, 1-Cbz-4-Piperidone, Benzyl 4-oxo-1-piperidinecarboxylate
Empirical Formula (Hill Notation):
C13H15NO3
CAS Number:
Molecular Weight:
233.26

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing