Skip to Content
MilliporeSigma
1-Thio-β-D-glucose tetraacetate 97%

1-Thio-β-D-glucose tetraacetate

1-Thio-β-D-glucose tetraacetate 97%
Synonym(s):
2,3,4,6-Tetra-O-acetyl-1-thio-β-D-glucopyranose
Empirical Formula (Hill Notation):
C14H20O9S
CAS Number:
Molecular Weight:
364.37
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing