Skip to Content
MilliporeSigma
(1S)-(−)-Verbenone analytical standard

(1S)-(−)-Verbenone

(1S)-(−)-Verbenone analytical standard
Synonym(s):
(1S,5S)-2-Pinen-4-one, (1S,5S)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-one
Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing