Skip to Content
MilliporeSigma
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 99%

(1S,2R)-(+)-2-Amino-1,2-diphenylethanol

(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 99%
Linear Formula:
C6H5CH(NH2)CH(C6H5)OH
CAS Number:
Molecular Weight:
213.28

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing