Skip to Content
MilliporeSigma
(1R)-(−)-Myrtenal ≥97%

(1R)-(−)-Myrtenal

(1R)-(−)-Myrtenal ≥97%
Synonym(s):
(−)-Myrtenal, (1R)-2-Pinen-10-al, (1R)-6,6-Dimethylbicyclo[3.1.1]hept-2-en-2-carboxaldehyde
Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing