Skip to Content
MilliporeSigma
(1R,2S)-(+)-cis-1-Amino-2-indanol 99%

(1R,2S)-(+)-cis-1-Amino-2-indanol

(1R,2S)-(+)-cis-1-Amino-2-indanol 99%
Synonym(s):
(1R,2S)-(+)-cis-1-Amino-2-hydroxyindane
Empirical Formula (Hill Notation):
C9H11NO
CAS Number:
Molecular Weight:
149.19

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing