Skip to Content
MilliporeSigma
1-O-Palmityl-sn-glycero-3-phosphocholine ≥99%, synthetic

1-O-Palmityl-sn-glycero-3-phosphocholine

1-O-Palmityl-sn-glycero-3-phosphocholine ≥99%, synthetic
Synonym(s):
1-Hexadecyl-sn-glycero-3-phosphocholine, 3-sn-Lysophosphatidylcholine, 1-hexadecyl, Lyso-PAF-C16, Lyso-platelet activating factor
Empirical Formula (Hill Notation):
C24H52NO6P
CAS Number:
Molecular Weight:
481.65

Products

Product Category
Brand
Biological Source
Formula Weight
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing