Skip to Content
MilliporeSigma
1-Methyladenosine ≥95% (HPLC)

1-Methyladenosine

1-Methyladenosine ≥95% (HPLC)
Synonym(s):
1-Methyladenosine, N1-Methyladenosine
Empirical Formula (Hill Notation):
C11H15N5O4
CAS Number:
Molecular Weight:
281.27

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing