Skip to Content
MilliporeSigma
1-Hydroxybenzotriazole hydrate wetted with not less than 20 wt. % water, 97% dry basis

1-Hydroxybenzotriazole hydrate

1-Hydroxybenzotriazole hydrate wetted with not less than 20 wt. % water, 97% dry basis
Synonym(s):
HOBt Hydrate
Empirical Formula (Hill Notation):
C6H5N3O · xH2O
CAS Number:
Molecular Weight:
135.12 (anhydrous basis)
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Application
Reaction Type