Skip to Content
MilliporeSigma
1-Ethyl-3-methylimidazolium dicyanamide ≥98.5% (HPLC/T)

1-Ethyl-3-methylimidazolium dicyanamide

1-Ethyl-3-methylimidazolium dicyanamide ≥98.5% (HPLC/T)
Synonym(s):
EMIM DCA
Empirical Formula (Hill Notation):
C8H11N5
CAS Number:
Molecular Weight:
177.21

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2