Skip to Content
MilliporeSigma
(1-Ethoxycyclopropoxy)trimethylsilane 98%

(1-Ethoxycyclopropoxy)trimethylsilane

(1-Ethoxycyclopropoxy)trimethylsilane 98%
Synonym(s):
Cyclopropanone ethyl trimethylsilyl acetal
Linear Formula:
C2H5OC3H4OSi(CH3)3
CAS Number:
Molecular Weight:
174.31

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing