Skip to Content
MilliporeSigma
1-Chloro-4-ethynylbenzene 98%

1-Chloro-4-ethynylbenzene

1-Chloro-4-ethynylbenzene 98%
Synonym(s):
(4-Chlorophenyl)acetylene
Linear Formula:
ClC6H4C≡CH
CAS Number:
Molecular Weight:
136.58

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing