Skip to Content
MilliporeSigma
1-Butanethiol ≥98%

1-Butanethiol

1-Butanethiol ≥98%
Synonym(s):
Butyl mercaptan, Mercaptan C4, 1-Butanethiol
Linear Formula:
CH3(CH2)3SH
CAS Number:
Molecular Weight:
90.19
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing