Skip to Content
MilliporeSigma
(1-Bromovinyl)trimethylsilane 97%

(1-Bromovinyl)trimethylsilane

(1-Bromovinyl)trimethylsilane 97%
Synonym(s):
α-(Trimethylsilyl)vinyl bromide
Linear Formula:
H2C=C(Br)Si(CH3)3
CAS Number:
Molecular Weight:
179.13
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing