Skip to Content
MilliporeSigma
1-Bromo-2-naphthoic acid ≥98%

1-Bromo-2-naphthoic acid

1-Bromo-2-naphthoic acid ≥98%
Synonym(s):
1-Bromo-2-naphthalenecarboxylic acid
Empirical Formula (Hill Notation):
C11H7BrO2
CAS Number:
Molecular Weight:
251.08
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing