Skip to Content
MilliporeSigma
1-(4-Methoxyphenyl)piperazine 97%

1-(4-Methoxyphenyl)piperazine

1-(4-Methoxyphenyl)piperazine 97%
Synonym(s):
1-(4-Anisyl)piperazine
Empirical Formula (Hill Notation):
C11H16N2O
CAS Number:
Molecular Weight:
192.26
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing