Skip to Content
MilliporeSigma
1,4-Diisocyanatobutane 97.0%

1,4-Diisocyanatobutane

1,4-Diisocyanatobutane 97.0%
Synonym(s):
Tetramethylene diisocyanate
Linear Formula:
OCN(CH2)4NCO
CAS Number:
Molecular Weight:
140.14

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing