Skip to Content
MilliporeSigma
1,4-Difluorobenzene analytical standard

1,4-Difluorobenzene

1,4-Difluorobenzene analytical standard
Empirical Formula (Hill Notation):
C6H4F2
CAS Number:
Molecular Weight:
114.09
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing