1,4,10,13-Tetraoxa-7,16-diazacyclooctadecane
Synonym(s):
1,10-Diaza-18-crown-6
Empirical Formula (Hill Notation):
C12H26N2O4
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing