Skip to Content
MilliporeSigma
1,3-Dinitrobenzene analytical standard

1,3-Dinitrobenzene

1,3-Dinitrobenzene analytical standard
Linear Formula:
C6H4(NO2)2
CAS Number:
Molecular Weight:
168.11
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing