Skip to Content
MilliporeSigma
1,3-Dimethoxybenzene ≥98%

1,3-Dimethoxybenzene

1,3-Dimethoxybenzene ≥98%
Synonym(s):
Dimethylresorcinol, Resorcinol dimethyl ether
Linear Formula:
C6H4(OCH3)2
CAS Number:
Molecular Weight:
138.16
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing