Skip to Content
MilliporeSigma
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane 97%

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane 97%
Synonym(s):
TIPDSiCl2
Linear Formula:
[(CH3)2CH]2Si(Cl)OSi(Cl)[CH(CH3)2]2
CAS Number:
Molecular Weight:
315.43

Products

Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing