Skip to Content
MilliporeSigma
1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine monohydrochloride 97%

1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine monohydrochloride

1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine monohydrochloride 97%
Empirical Formula (Hill Notation):
C13H18Cl2N2 · HCl
CAS Number:
Molecular Weight:
309.66
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing