Skip to Content
MilliporeSigma
1,2-Bis(dimethylphosphino)ethane 97%

1,2-Bis(dimethylphosphino)ethane

1,2-Bis(dimethylphosphino)ethane 97%
Synonym(s):
DMPE, Dimethyl-2-(dimethylphosphino)ethylphosphine, Ethylenebis(dimethylphosphine)
Linear Formula:
(CH3)2PCH2CH2P(CH3)2
CAS Number:
Molecular Weight:
150.14

Products

Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing