MilliporeSigma
Azides
A white plastic bottle with a red cap labeled “Sodium azide ReagentPlus®, 99.5%”. On the label, there are hazard symbols indicating flammability, an exclamation mark denoting a warning, and a health hazard icon. Below the symbols is a red diamond-shaped label with a fire symbol indicating its flammability and danger if it catches fire. The product code “1006” is visible at the top of the label.
Azides are chemical compounds characterized by the presence of an azido group, which is a linear arrangement of three nitrogen atoms. Azides originate from hydrazoic acid (HN3), which can take the form of an inorganic salt like sodium azide (NaN3). Alternatively, they can be derived from an organic compound wherein the hydrogen atom of hydrazoic acid is substituted. This substitution can involve a hydrocarbon group, as seen in alkyl or aryl azide (RN3), or an acyl (carboxylic acid) group, as observed in acyl azide.
These energy-rich molecules find numerous applications. For instance, sodium azide serves as a preservative, mutagen, biocide, and assay reagent. Organic azides, on the other hand, exhibit a wide range of organic reactions and play a crucial role in Click Chemistry. It is essential to exercise caution when working with azides since both organic and inorganic variants can be heat and shock-sensitive, potentially undergoing explosive decomposition with minimal external energy input.
Read more about
Products
Compare
Product Number
Product Name
Product Description
Pricing
Azides as Reagents in Click Chemistry
Organic azides play a crucial role in various organic reactions, notably as key components in the azide-alkyne "click" reaction, commonly achieved via the Cu(I)-catalyzed Huisgen azide-alkyne 1,3-dipolar cycloaddition. Since the pioneering synthesis of the first organic azide, phenyl azide, by Peter Griess in 1864, these versatile compounds have sparked significant interest. They find extensive applications in combinatorial synthesis, peptide and heterocycle synthesis, and the modification of biopolymers. Prominent applications include azide-alkyne cycloadditions and the Staudinger ligation in various forms. The azido group proves valuable as a protecting group for primary amines, especially in sensitive substrates like complex carbohydrates, peptide nucleic acids (PNA), and coordination compounds, thanks to its stability under alkene metathesis conditions. Incorporating azido functional groups into organic molecules holds increasing importance, significantly impacting both the fields of organic chemistry and biology, from amino group protection to chemical ligation. A variety of azide sources, ranging from sodium azide to diphenyl phosphoryl azide, are available to facilitate azide synthesis and the preparation of customized organic azides.
Sodium Azide
Sodium azide (NaN3) is an inorganic compound known for its potent inhibitory properties. It exists in the form of a water-soluble crystalline powder with no discernible odor. This ionic substance is widely recognized for its versatile applications in cell culture, molecular biology, and biochemical research. Frequently used as a bacteriostatic preservative in water-based lab reagents and biological fluids, sodium azide acts as a metabolic inhibitor, disrupting oxidative phosphorylation. In cell culture, it plays a crucial role in maintaining cell line integrity by preventing microbial contamination. Furthermore, sodium azide finds application in molecular biology, where it is used to preserve nucleic acids, ensuring the accuracy of results in subsequent analyses. Beyond these applications, sodium azide serves diverse purposes in different fields. It transforms Baylis-Hillman acetates into ethyl (E)-2-azidomethyl-3-phenylpropenoate in aqueous conditions. In histopathology, it is used to prepare and store tissue samples, and it is also utilized as a component of the staining buffer for whole-mount immunolabeling. Additionally, sodium azide acts as a catalyst for oxidative decarboxylation and Michael addition reactions. It serves as a reagent for synthesizing various compounds, including blue fluorescent copolymers, metal phosphonates, and arenes through aminations. Additionally, sodium azide acts as a catalyst for oxidative decarboxylation and Michael addition reactions. In clinical settings, sodium azide is utilized as a preservative in diluting fluid used for red blood cell counting, and it prevents capping and internalization of fluorescent surface-bound antibodies in scientific applications like flow cytometry.