Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1A01260

USP

(16a)-16,17,21-Trihydroxypregna-1,4,9(11)-triene-3,20-dione

Pharmaceutical Analytical Impurity (PAI)

Synonym(s):

(16α)-16,17,21-Trihydroxypregna-1,4,9(11)-triene-3,20-dione, (8S,10S,13S,14S,16R,17S)-16,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,10,12,13,14,15,16,17-decahydro-3H-cyclopenta[a]phenanthren-3-one, 16α,17,21-Trihydroxy-pregna-1,4,9(11)-triene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O5
CAS Number:
Molecular Weight:
358.43
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical analytical impurity (PAI)

Agency

USP

API family

budesonide

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

(16α)-16,17,21-Trihydroxypregna-1,4,9(11)-triene-3,20-dione is a USP Pharmaceutical Analytical Impurity (PAI).

USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.

Associated Drug Substance: Budesonide

Therapeutic Area: Steroids

For more information about this PAI, visit here.

Application

(16α)-16,17,21-Trihydroxypregna-1,4,9(11)-triene-3,20-dione (USP PAI) is intended for use in analytical testing to detect, identify, and measure pharmaceutical impurities.

Features and Benefits

USP PAI advance your early analytical R&D and process development. PAI can be used in the following applications:

1. Conduct analytical tests during early formulation feasibility studies.
2. Determine degradation impurities produced during stress studies.
3. Develop, validate, and transfer analytical methods.
4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.
5. Record retention times and/or spectra
6. Determine relative response factors.
7. Identify unknown impurities that formed during ICH stability conditions.
8. Identify impurities that are present in the Reference Listed Drug
9. Test for and profile impurities not listed in drug substance and drug product monographs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service