Skip to Content
MilliporeSigma

Skip To

1614909

USP

Sodium tartrate dihydrate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Sodium tartrate dibasic dihydrate, L-(+)-Tartaric acid disodium salt, Disodium tartrate dihydrate, Sodium tartrate dihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
4 x 500 mg

Estimated to ship TODAYfromMILWAUKEE

$276.00

About This Item

Empirical Formula (Hill Notation):
C4H4Na2O6 · 2H2O
CAS Number:
Molecular Weight:
230.08
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
6121732

$276.00


Estimated to ship TODAYDetails


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

sodium tartrate dihydrate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[Na+].[H]O[H].[H]O[H].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.2Na.2H2O/c5-1(3(7)8)2(6)4(9)10;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;2*1H2/q;2*+1;;/p-2/t1-,2-;;;;/m1..../s1

InChI key

FGJLAJMGHXGFDE-DGFHWNFOSA-L

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium tartrate dihydrate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
719951.93250S4797
grade

pharmaceutical primary standard

grade

-

grade

ACS reagent

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

format

neat

format

-

format

-

format

-

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

application(s)

-

API family

sodium tartrate dihydrate

API family

-

API family

-

API family

-


Still not finding the right product?

Explore all of our products under Sodium tartrate dihydrate


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters