Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0002399

Supelco

Fluocortolone pivalate for peak identification CRS

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Fluocortolone pivalate, (6α,11β,16α)-21-(2,2-Dimethyl-1-oxopropoxy)-6-fluoro-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H37FO5
CAS Number:
Molecular Weight:
460.58
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

F[C@H]1C[C@@H]2[C@@H]([C@@]4(C1=CC(=O)C=C4)C)[C@H](C[C@]3([C@H]2C[C@H]([C@@H]3C(=O)COC(=O)C(C)(C)C)C)C)O

InChI

1S/C27H37FO5/c1-14-9-17-16-11-19(28)18-10-15(29)7-8-26(18,5)23(16)20(30)12-27(17,6)22(14)21(31)13-33-24(32)25(2,3)4/h7-8,10,14,16-17,19-20,22-23,30H,9,11-13H2,1-6H3/t14-,16+,17+,19+,20+,22-,23-,26+,27+/m1/s1

InChI key

XZBJVIQXJHGUBE-HZMVJJPJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. Compendial materials should be used for the purpose described in the appropriate monograph unless otherwise authorized by the National Competent Authority. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

e.g. pharmaceutical (small molecule)

Packaging

Unit quantity: 10mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service