Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL89220

α-Hederin

phyproof® Reference Substance

Synonym(s):

(3β,4α)-3-[[2-O-(6-Deoxy-α-L-mannopyranosyl)-α-L-arabinopyranosyl]oxy]-23-hydroxyolean-12-en-28-oic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H66O12
CAS Number:
Molecular Weight:
750.96
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@@H](O)CO[C@H]2O[C@H]3CC[C@@]4(C)[C@@H](CC[C@]5(C)[C@@H]4CC=C6[C@@H]7CC(C)(C)CC[C@@]7(CC[C@@]56C)C(O)=O)[C@]3(C)CO)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C41H66O12/c1-21-28(44)30(46)31(47)33(51-21)53-32-29(45)24(43)19-50-34(32)52-27-11-12-37(4)25(38(27,5)20-42)10-13-40(7)26(37)9-8-22-23-18-36(2,3)14-16-41(23,35(48)49)17-15-39(22,40)6/h8,21,23-34,42-47H,9-20H2,1-7H3,(H,48,49)/t21-,23-,24-,25+,26+,27-,28-,29-,30+,31+,32+,33-,34-,37-,38-,39+,40+,41-/m0/s1

InChI key

KEOITPILCOILGM-LLJOFIFVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application

  • α-Hederin as a potent inhibitor in cancer research: α-Hederin has been identified and validated as a potent inhibitor of the Wnt/β-catenin signaling pathway in breast cancer stem cells, offering a promising avenue for targeted cancer therapy and highlighting its role in anti-cancer pharmacological research (Saliu et al., 2024).
  • α-Hederin in overcoming drug resistance: This natural triterpenoid saponin has shown efficacy in reprogramming miRNA activity to overcome paclitaxel resistance mediated by small extracellular vesicles in non-small cell lung cancer (NSCLC), reinforcing its potential as a valuable agent in respiratory research and cancer treatment (Chang et al., 2024).
  • α-Hederin′s role in promoting ferroptosis in cancer: Research indicates that α-Hederin can induce ferroptosis and reverse chemoresistance to cisplatin in non-small cell lung cancer, providing insights into its mechanism as a cytotoxic agent and its applications in enhancing the efficacy of conventional chemotherapies (Han et al., 2024).
  • Molecular mechanisms of α-Hederin in tumor progression: A review of the molecular mechanisms by which α-Hederin influences tumor progression highlights its potential uses in biochemistry and pharmaceutical research, particularly for its anti-inflammatory and anticancer properties (Meng et al., 2024).

Other Notes

This compound is commonly found in plants of the genus: hedera

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service