Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BP1078

5-Methoxytryptamine

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

2-(5-Methoxyindol-3-yl)ethylamine, 3-(2-Aminoethyl)-5-methoxyindole, 5-MOT, 5-Methoxyindole-3-ethanamine, Deacetylmelatonin, NSC 56422

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O
CAS Number:
Molecular Weight:
190.24
Beilstein:
145587
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

melatonin

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

121-123 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc2[nH]cc(CCN)c2c1

InChI

1S/C11H14N2O/c1-14-9-2-3-11-10(6-9)8(4-5-12)7-13-11/h2-3,6-7,13H,4-5,12H2,1H3

InChI key

JTEJPPKMYBDEMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

5-Methoxytryptamine BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Melatonin Capsules
  • Reactant for preparation of:
    • Carboline disaccharide domain of shishijimicin A
    • Melatonin analogs for the reduction of intraocular pressure
    • 5-HT4 receptor ligands
    • inhibitors of sortase A and isocitrate lyase
    • Therapeutic agents for treatment of ischemia/reperfusion (I/R) injury
    • Aurora and epidermal growth factor receptor kinase inhibitors
    • Agents for the treatment of human papillomavirus infection
    • Manzamine analogues for the control of neuroinflammation and cerebral infections
    • Inhibitors of pro-inflammatory cytokines
    • Tacrine-melatonin hybrids as multifunctional agents for alzheimer′s disease

    Packaging

    Unit quantity: 25 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Pictograms

    Exclamation mark

    Signal Word

    Warning

    Hazard Statements

    Precautionary Statements

    Hazard Classifications

    Acute Tox. 4 Oral

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service