Skip to Content
MilliporeSigma
All Photos(1)

Documents

89471

Supelco

Heavy metal mix IX

(in 12% nitric acid), TraceCERT®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

composition

As, 100 mg/L
Cd, 100 mg/L
Hg, 100 mg/L
Pb, 100 mg/L

concentration

(in 12% nitric acid)

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

multi-component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ICP-MS analyzes 12 heavy metals in hemp-containing food samples using standard addition calibration for accurate quantification.

ICP-MS analyzes 12 heavy metals in hemp-containing food samples using standard addition calibration for accurate quantification.

ICP-MS analyzes 12 heavy metals in hemp-containing food samples using standard addition calibration for accurate quantification.

ICP-MS analyzes 12 heavy metals in hemp-containing food samples using standard addition calibration for accurate quantification.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service