Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69085

Supelco

Di(ethylene glycol) dibenzoate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CO2CH2CH2)2O
CAS Number:
Molecular Weight:
314.33
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Assay

≥95%

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.544 (lit.)

bp

235-237 °C/7 mmHg (lit.)

density

1.175 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

O=C(OCCOCCOC(=O)c1ccccc1)c2ccccc2

InChI

1S/C18H18O5/c19-17(15-7-3-1-4-8-15)22-13-11-21-12-14-23-18(20)16-9-5-2-6-10-16/h1-10H,11-14H2

InChI key

NXQMCAOPTPLPRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service