Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67283-U

Supelco

BIOshell A160 Peptide C18 (2 µm) HPLC Columns

L × I.D. 15 cm × 1.5 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

product name

BIOshell A160 Peptide C18 Column, 2 µm HPLC Column, 2.0 μm particle size, L × I.D. 15 cm × 1.5 mm

material

stainless steel hardware

Quality Level

Agency

suitable for USP L1

product line

BIOshell

feature

endcapped: no

analyte chemical class(es)

peptides

manufacturer/tradename

BIOshell

packaging

1 ea of

parameter

14,500 psi max. pressure (1000 bar)
60 °C max. temp.

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable

L × I.D.

15 cm × 1.5 mm

matrix

spherical silica particle platform
superficially porous particle

matrix active group

C18 (octadecyl) phase

particle size

2.0 μm

pore size

160 Å pore size

operating pH range

1-8

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Shell thickness: 0.4 μm
Solid Core: 1.2 μm
Extent of labeling: 4.0%

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service